missing translation for 'onlineSavingsMsg'
Learn More

N,N-Dimethylacetamide, 99%

Catalog No. AAA10924AP
Change view
Click to view available options
Quantity:
250 mL
500 mL
2500 mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA10924AP 500 mL
AAA10924AK 250 mL
AAA109240F 2500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA10924AP Supplier Thermo Scientific Chemicals Supplier No. A10924AP

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 127-19-5 | C4H9NO | 87.12 g/mol

N,N-Dimethylacetamide is used as a polar solvent in organic synthesis as well as in reactions involving strong bases such as sodium hydroxide. It is also used as a solvent for fibers like polyacrylonitrile, spandex, as an excipient in drugs viz. vumon, busulfex and in the adhesive industry. It finds application as a reaction medium in the production of pharmaceuticals, agrochemicals, dyes and plasticizers. It is widely used in polymer industry due to its high solving power for high molecular-weight polymers and synthetic resins. It plays an important role as a catalyst in various reactions viz. cyclization, halogenation, cynidation, alkylation and dehydrogenation and increases the yield of main products. It is also used as an extracting agent for oil and gases, in paint removers, in the production of photo-resist stripping compounds and as booster solvent in coatings.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
N,N-Dimethylacetamide is used as a polar solvent in organic synthesis as well as in reactions involving strong bases such as sodium hydroxide. It is also used as a solvent for fibers like polyacrylonitrile, spandex, as an excipient in drugs viz. vumon, busulfex and in the adhesive industry. It finds application as a reaction medium in the production of pharmaceuticals, agrochemicals, dyes and plasticizers. It is widely used in polymer industry due to its high solving power for high molecular-weight polymers and synthetic resins. It plays an important role as a catalyst in various reactions viz. cyclization, halogenation, cynidation, alkylation and dehydrogenation and increases the yield of main products. It is also used as an extracting agent for oil and gases, in paint removers, in the production of photo-resist stripping compounds and as booster solvent in coatings.

Solubility
Miscible with water, benzene, alcohol, acetone, ether, esters and ketones.

Notes
Hygroscopic. Incompatible with strong oxidizing agents.

Chemical Identifiers

CAS 127-19-5
Molecular Formula C4H9NO
Molecular Weight (g/mol) 87.12
MDL Number MFCD00008686
InChI Key FXHOOIRPVKKKFG-UHFFFAOYSA-N
Synonym dimethylacetamide, dmac, acetamide, n,n-dimethyl, acetdimethylamide, dimethyl acetamide, n,n-dimethyl acetamide, dimethylamide acetate, n,n-dimethylethanamide, dimethylacetone amide, acetyldimethylamine
PubChem CID 31374
ChEBI CHEBI:84254
IUPAC Name N,N-dimethylacetamide
SMILES CN(C)C(C)=O

Specifications

Melting Point -20°C
Density 0.94
Boiling Point 165°C to 166°C
Flash Point 70°C (158°F)
Odor Ammonia-like
Refractive Index 1.4385
Linear Formula CH3CON(CH3)2
Quantity 500 mL
Beilstein 1737614
Merck Index 14,3227
Solubility Information Miscible with water,benzene,alcohol,acetone,ether,esters and ketones.
Formula Weight 87.12
Percent Purity 99%
Chemical Name or Material N,N-Dimethylacetamide
Show More Show Less
Hazard Category H227-H312+H332-H360D
Hazard Statement GHS H Statement
H360-H312-H332-H227
May damage fertility or the unborn child.
Harmful in contact with skin.
Harmful if inhaled.
Combustible liquid.
Precautionary Statement P201-P202-P210-P261-P271-P281-P302+P352-P304+P340-P308+P313-P312-P363-P370+P378q-P501c
DOTInformation Hazard Class: 6.1; Packaging Group: III
EINECSNumber 204-826-4
RTECSNumber AB7700000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only