missing translation for 'onlineSavingsMsg'
Learn More
Learn More
N,N-Dimethylacetamide, ≥99.5% (GC), Honeywell™ Riedel-de-Haën™
Puriss. p.a.,≥99.5% (GC)
$507.63 - $507.63
Chemical Identifiers
| CAS | 127-19-5 |
|---|---|
| Molecular Formula | C4H9NO |
| Molecular Weight (g/mol) | 87.12 |
| MDL Number | MFCD00008686 |
| InChI Key | FXHOOIRPVKKKFG-UHFFFAOYSA-N |
| Synonym | dimethylacetamide, dmac, acetamide, n,n-dimethyl, acetdimethylamide, dimethyl acetamide, n,n-dimethyl acetamide, dimethylamide acetate, n,n-dimethylethanamide, dimethylacetone amide, acetyldimethylamine |
| PubChem CID | 31374 |
| ChEBI | CHEBI:84254 |
| IUPAC Name | N,N-dimethylacetamide |
| SMILES | CN(C)C(C)=O |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
60-048-506
|
Honeywell Inc
388401L |
1 L |
Each for $507.63
|
|
|||||
Description
- Honeywell Riedel-de Haen has over 200 years of history in producing high-purity chemicals
- It is the leading brand offering High-Purity Solvents for dedicated applications.
Chemical Identifiers
| 127-19-5 | |
| 87.12 | |
| FXHOOIRPVKKKFG-UHFFFAOYSA-N | |
| 31374 | |
| N,N-dimethylacetamide |
| C4H9NO | |
| MFCD00008686 | |
| dimethylacetamide, dmac, acetamide, n,n-dimethyl, acetdimethylamide, dimethyl acetamide, n,n-dimethyl acetamide, dimethylamide acetate, n,n-dimethylethanamide, dimethylacetone amide, acetyldimethylamine | |
| CHEBI:84254 | |
| CN(C)C(C)=O |
Specifications
| 127-19-5 | |
| 1 L | |
| 70°C (158°F) | |
| C4H9NO | |
| MFCD00008686 | |
| 1737614 | |
| FXHOOIRPVKKKFG-UHFFFAOYSA-N | |
| N,N-dimethylacetamide | |
| 31374 | |
| 87.12g/mol | |
| N,N-Dimethylacetamide |
| 0.937g/mL at 25°C | |
| 1.8% (100°F), 11.5% (160°F) | |
| 914°F | |
| CH3CON(CH3)2 | |
| 4mmHg (38°C), 2mmHg (25°C) | |
| dimethylacetamide, dmac, acetamide, n,n-dimethyl, acetdimethylamide, dimethyl acetamide, n,n-dimethyl acetamide, dimethylamide acetate, n,n-dimethylethanamide, dimethylacetone amide, acetyldimethylamine | |
| CN(C)C(C)=O | |
| 87.12 | |
| CHEBI:84254 | |
| ≥99.5% (GC) |
Safety and Handling
P201-P261-P280-P305 + P351 + P338-P308 + P313
RTECSNumber : 204-826-4