missing translation for 'onlineSavingsMsg'
Learn More

N,N-Dimethylacetamide, 98% min., MilliporeSigma™

Catalog No. MDX15453
Change view
Click to view available options
Quantity:
4 L
200 L
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MDX15453 4 L
MDX154530 200 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. MDX15453 Supplier Technidata Supplier No. DX15453

Chemical Identifiers

CAS 127-19-5
Molecular Formula C4H9NO
Molecular Weight (g/mol) 87.12
MDL Number MFCD00008686
InChI Key FXHOOIRPVKKKFG-UHFFFAOYSA-N
Synonym dimethylacetamide, dmac, acetamide, n,n-dimethyl, acetdimethylamide, dimethyl acetamide, n,n-dimethyl acetamide, dimethylamide acetate, n,n-dimethylethanamide, dimethylacetone amide, acetyldimethylamine
PubChem CID 31374
ChEBI CHEBI:84254
IUPAC Name N,N-dimethylacetamide
SMILES CN(C)C(C)=O

Specifications

Color Colorless
Quantity 4 L
Physical Form Liquid
Assay Percent Range 98% Min.
Linear Formula CH3CON(CH3)2
Formula Weight 87.12
Chemical Name or Material N, N-Dimethylacetamide
Recommended Storage Room Temperature