missing translation for 'onlineSavingsMsg'
Learn More

N,N-Dimethyl-p-phenylenediamine sulfate, 98%

Catalog No. AAA1417536
Change view
Click to view available options
Quantity:
500 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1417536 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAA1417536 Supplier Thermo Scientific Chemicals Supplier No. A1417536
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 536-47-0 | C8H14N2O4S | 234.27 g/mol

N,N-Dimethyl-p-phenylenediamine sulfate salt is used to investigate the role of H2S in cardioprotection induced by ischemic preconditioning in rat heart, in spectrophotometric determination of gold(III) in gold-pharmaceuticals such as sodium aurothimaleate and auranofin, to investigate colonic tissue H2S production using the acetate trapping assay system.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
N,N-Dimethyl-p-phenylenediamine sulfate salt is used to investigate the role of H2S in cardioprotection induced by ischemic preconditioning in rat heart, in spectrophotometric determination of gold(III) in gold-pharmaceuticals such as sodium aurothimaleate and auranofin, to investigate colonic tissue H2S production using the acetate trapping assay system.

Solubility
Soluble in water.

Notes
Light Sensitive & Hygroscopic. Store in the dark. Incompatible with oxidizing agents and light.

Chemical Identifiers

CAS 536-47-0
Molecular Formula C8H14N2O4S
Molecular Weight (g/mol) 234.27
MDL Number MFCD00012992
InChI Key GLUKPDKNLKRLHX-UHFFFAOYSA-N
Synonym n,n-dimethyl-1,4-phenylenediamine sulfate, n,n-dimethyl-p-phenylenediamine sulfate, n1,n1-dimethylbenzene-1,4-diamine sulfate, 1,4-benzenediamine, n,n-dimethyl-, sulfate, unii-x997ivp3ja, 4-amino-n,n-dimethylaniline sulphate, x997ivp3ja, 1,4-benzenediamine, n,n-dimethyl-, sulfate 1:1, dimethyl-p-phenylenediamine; sulfuric acid, 1,4-benzenediamine, n1,n1-dimethyl-, sulfate 1:?
PubChem CID 80351
IUPAC Name 4-N,4-N-dimethylbenzene-1,4-diamine;sulfuric acid
SMILES OS(O)(=O)=O.CN(C)C1=CC=C(N)C=C1

Specifications

Melting Point 200°C to 205°C (decomposition)
Odor Odorless
Quantity 500 g
UN Number UN2811
Beilstein 4612278
Sensitivity Hygroscopic; Light sensitive
Solubility Information Soluble in water.
Formula Weight 234.28
Percent Purity 98%
Chemical Name or Material N,N-Dimethyl-p-phenylenediamine sulfate
Hazard Category H301+H311+H331-H315-H319-H335
Hazard Statement GHS H Statement
H301-H310-H331-H315-H319-H335
Toxic if swallowed.
Fatal in contact with skin.
Toxic if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P310-P302+P352-P304+P340-P305+P351+P338-P311-P312-P330-P332+P313-P361-P363-P501c
DOTInformation Hazard Class: 6.1; Packaging Group: I
EINECSNumber 208-636-2
RTECSNumber ST1575000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only