missing translation for 'onlineSavingsMsg'
Learn More

N,N-Dimethyl-n-octadecylamine 90.0+%, TCI America™

Catalog No. D160925G
Change view
Click to view available options
Quantity:
25 g
250 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D160925G 25 g
D1609250G 250 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D160925G Supplier TCI America Supplier No. D160925G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 124-28-7
Molecular Formula C20H44ClN
Molecular Weight (g/mol) 334.03
MDL Number MFCD00048496
InChI Key PFKRTWCFCOUBHS-UHFFFAOYSA-N
Synonym n,n-dimethyloctadecylamine, dymanthine, dimantine, n,n-dimethyl-n-octadecylamine, dimethyloctadecylamine, 1-octadecanamine, n,n-dimethyl, n,n-dimethyl-1-octadecanamine, dimethyl stearamine, stearyldimethylamine, n,n-dimethylstearylamine
PubChem CID 15365
IUPAC Name dimethyl(octadecyl)azanium chloride
SMILES [Cl-].CCCCCCCCCCCCCCCCCC[NH+](C)C

Specifications

Color White
Quantity 25 g
UN Number 3259
Formula Weight 297.57
Percent Purity ≥90.0% (GC)
Grade Not Available
Physical Form Crystalline Lumps
Chemical Name or Material N,N-Dimethyl-n-octadecylamine
EINECSNumber (2)-0176&(2)-0185
RTECSNumber RG4200000
TSCA Yes