missing translation for 'onlineSavingsMsg'
Learn More
Learn More
N,N-Dimethyl-N-(methylsulfanylmethylene)ammonium Iodide 98.0+%, TCI America™
Supplier: TCI America D434025G
Specifications
| N,N-Dimethyl-N-(methylsulfanylmethylene)ammonium Iodide | |
| 125°C | |
| C4H10INS | |
| 25 g | |
| BPBAGFOLLWBKGJ-UHFFFAOYSA-M | |
| dimethyl[(methylsulfanyl)methylidene]azanium iodide | |
| 12347574 | |
| ≥98.0% (T) |
| 29085-13-0 | |
| White | |
| MFCD17015287 | |
| N,N-Dimethyl-N-(methylsulfanylmethylene)iminium Iodide | |
| [I-].CSC=[N+](C)C | |
| 231.10 | |
| 231.10 | |
| Crystalline Powder |
Chemical Identifiers
| 29085-13-0 | |
| 231.10 | |
| BPBAGFOLLWBKGJ-UHFFFAOYSA-M | |
| 12347574 | |
| [I-].CSC=[N+](C)C |
| C4H10INS | |
| MFCD17015287 | |
| N,N-Dimethyl-N-(methylsulfanylmethylene)iminium Iodide | |
| dimethyl[(methylsulfanyl)methylidene]azanium iodide |
Safety and Handling
TSCA : No
Recommended Storage : Refrigerator