missing translation for 'onlineSavingsMsg'
Learn More

N,N-Dimethyl-N-(methylsulfanylmethylene)ammonium Iodide 98.0+%, TCI America™

Catalog No. D43405G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D43405G 5 g
D434025G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D43405G Supplier TCI America Supplier No. D43405G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 29085-13-0
Molecular Formula C4H10INS
Molecular Weight (g/mol) 231.10
MDL Number MFCD17015287
InChI Key BPBAGFOLLWBKGJ-UHFFFAOYSA-M
Synonym N,N-Dimethyl-N-(methylsulfanylmethylene)iminium Iodide
PubChem CID 12347574
IUPAC Name dimethyl[(methylsulfanyl)methylidene]azanium iodide
SMILES [I-].CSC=[N+](C)C

Specifications

Melting Point 125°C
Color White
Quantity 5 g
Formula Weight 231.10
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material N,N-Dimethyl-N-(methylsulfanylmethylene)ammonium Iodide
TSCA No
Recommended Storage Refrigerator