missing translation for 'onlineSavingsMsg'
Learn More

N,N-Dimethyl-L-phenylalanine, 99%

CAS: 17469-89-5 | C11H15NO2 | 193.25 g/mol

Supplier:  Thermo Scientific Chemicals 440030050

 View more versions of this product

Catalog No. AC440030050


Only null left
Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

N, N-Dimethyl-L-phenylalanine
98.5
225.0°C to 227.0°C
0.0950g/mL
98.5% min. (HPLC)
C11H15NO2
5 g
73.92
Solubility in water: in water:(20°C) freely soluble
CN(C)[C@@H](CC1=CC=CC=C1)C(O)=O
193.25
193.25
Powder
17469-89-5
100.0
White
Authentic
Glass bottle
MFCD00082539
0.095
n,n-dimethyl-l-phenylalanine, phenylalanine, n,n-dimethyl, n,n-dimethylphenylalanine, l-phenylalanine, n,n-dimethyl, s-2-dimethylamino-3-phenylpropanoic acid, 2s-2-dimethylamino-3-phenylpropanoic acid, alanine, n,n-dimethyl-3-phenyl-, l, l-phenylalanine,n,n-dimethyl, s-2-dimethylamino-3-phenyl-propionic acid
HOGIQTACRLIOHC-JTQLQIEISA-N
(2S)-2-(dimethylamino)-3-phenylpropanoic acid
11830289
99%

Chemical Identifiers

17469-89-5
193.25
HOGIQTACRLIOHC-JTQLQIEISA-N
11830289
CN(C)[C@@H](CC1=CC=CC=C1)C(O)=O
C11H15NO2
MFCD00082539
n,n-dimethyl-l-phenylalanine, phenylalanine, n,n-dimethyl, n,n-dimethylphenylalanine, l-phenylalanine, n,n-dimethyl, s-2-dimethylamino-3-phenylpropanoic acid, 2s-2-dimethylamino-3-phenylpropanoic acid, alanine, n,n-dimethyl-3-phenyl-, l, l-phenylalanine,n,n-dimethyl, s-2-dimethylamino-3-phenyl-propionic acid
(2S)-2-(dimethylamino)-3-phenylpropanoic acid
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only