missing translation for 'onlineSavingsMsg'
Learn More
Learn More
N,N-Dimethyl-4-nitrosoaniline 98.0+%, TCI America™
$137.94 - $1610.54
Chemical Identifiers
| CAS | 138-89-6 |
|---|---|
| Molecular Formula | C8H10N2O |
| Molecular Weight (g/mol) | 150.181 |
| MDL Number | MFCD00002063 |
| InChI Key | CMEWLCATCRTSGF-UHFFFAOYSA-N |
| Synonym | ndma, accelerine, p-nitroso-n,n-dimethylaniline, n,n-dimethyl-p-nitrosoaniline, 4-nitroso-n,n-dimethylaniline, 4-nitrosodimethylaniline, p-nitrosodimethylanilide, paranitrosodimethylanilide, 4-dimethylamino nitrosobenzene, benzenamine, n,n-dimethyl-4-nitroso |
| PubChem CID | 8749 |
| ChEBI | CHEBI:59990 |
| IUPAC Name | N,N-dimethyl-4-nitrosoaniline |
| SMILES | CN(C)C1=CC=C(C=C1)N=O |
Chemical Identifiers
| 138-89-6 | |
| 150.181 | |
| CMEWLCATCRTSGF-UHFFFAOYSA-N | |
| 8749 | |
| N,N-dimethyl-4-nitrosoaniline |
| C8H10N2O | |
| MFCD00002063 | |
| ndma, accelerine, p-nitroso-n,n-dimethylaniline, n,n-dimethyl-p-nitrosoaniline, 4-nitroso-n,n-dimethylaniline, 4-nitrosodimethylaniline, p-nitrosodimethylanilide, paranitrosodimethylanilide, 4-dimethylamino nitrosobenzene, benzenamine, n,n-dimethyl-4-nitroso | |
| CHEBI:59990 | |
| CN(C)C1=CC=C(C=C1)N=O |
Specifications
| 138-89-6 | |
| Green-Yellow | |
| MFCD00002063 | |
| 1369 | |
| CMEWLCATCRTSGF-UHFFFAOYSA-N | |
| N,N-dimethyl-4-nitrosoaniline | |
| 8749 | |
| 150.18 | |
| Crystalline Powder |
| 85°C | |
| C8H10N2O | |
| 25 g | |
| ndma, accelerine, p-nitroso-n,n-dimethylaniline, n,n-dimethyl-p-nitrosoaniline, 4-nitroso-n,n-dimethylaniline, 4-nitrosodimethylaniline, p-nitrosodimethylanilide, paranitrosodimethylanilide, 4-dimethylamino nitrosobenzene, benzenamine, n,n-dimethyl-4-nitroso | |
| CN(C)C1=CC=C(C=C1)N=O | |
| 150.181 | |
| CHEBI:59990 | |
| ≥98.0% (GC,T) | |
| N,N-Dimethyl-4-nitrosoaniline |
Safety and Handling
RTECSNumber : BX7175000
TSCA : Yes