missing translation for 'onlineSavingsMsg'
Learn More

N,N-Dimethyl-4-nitrosoaniline 98.0+%, TCI America™

Catalog No. N026225G
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N026225G 25 g
N0262500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. N026225G Supplier TCI America Supplier No. N026225G

Chemical Identifiers

CAS 138-89-6
Molecular Formula C8H10N2O
Molecular Weight (g/mol) 150.181
MDL Number MFCD00002063
InChI Key CMEWLCATCRTSGF-UHFFFAOYSA-N
Synonym ndma, accelerine, p-nitroso-n,n-dimethylaniline, n,n-dimethyl-p-nitrosoaniline, 4-nitroso-n,n-dimethylaniline, 4-nitrosodimethylaniline, p-nitrosodimethylanilide, paranitrosodimethylanilide, 4-dimethylamino nitrosobenzene, benzenamine, n,n-dimethyl-4-nitroso
PubChem CID 8749
ChEBI CHEBI:59990
IUPAC Name N,N-dimethyl-4-nitrosoaniline
SMILES CN(C)C1=CC=C(C=C1)N=O

Specifications

Melting Point 85°C
Color Green-Yellow
Quantity 25 g
UN Number 1369
Formula Weight 150.18
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material N,N-Dimethyl-4-nitrosoaniline
RTECSNumber BX7175000
TSCA Yes