Learn More
N,N-Dimethyl-4-nitrosoaniline, 98%
CAS: 138-89-6 | C8H10N2O | 150.181 g/mol
$82.81 - $436.03
Chemical Identifiers
| CAS | 138-89-6 |
|---|---|
| Molecular Formula | C8H10N2O |
| Molecular Weight (g/mol) | 150.181 |
| MDL Number | MFCD00002063 |
| InChI Key | CMEWLCATCRTSGF-UHFFFAOYSA-N |
| Synonym | ndma, accelerine, p-nitroso-n,n-dimethylaniline, n,n-dimethyl-p-nitrosoaniline, 4-nitroso-n,n-dimethylaniline, 4-nitrosodimethylaniline, p-nitrosodimethylanilide, paranitrosodimethylanilide, 4-dimethylamino nitrosobenzene, benzenamine, n,n-dimethyl-4-nitroso |
| PubChem CID | 8749 |
| ChEBI | CHEBI:59990 |
| IUPAC Name | N,N-dimethyl-4-nitrosoaniline |
| SMILES | CN(C)C1=CC=C(C=C1)N=O |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AAA1830114
|
Thermo Scientific Chemicals
A1830114 |
25 g |
Each for $82.81
|
|
|||||
|
AAA1830130
|
Thermo Scientific Chemicals
A1830130 |
250 g |
Each for $436.03
|
|
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 138-89-6 | |
| 150.181 | |
| CMEWLCATCRTSGF-UHFFFAOYSA-N | |
| 8749 | |
| N,N-dimethyl-4-nitrosoaniline |
| C8H10N2O | |
| MFCD00002063 | |
| ndma, accelerine, p-nitroso-n,n-dimethylaniline, n,n-dimethyl-p-nitrosoaniline, 4-nitroso-n,n-dimethylaniline, 4-nitrosodimethylaniline, p-nitrosodimethylanilide, paranitrosodimethylanilide, 4-dimethylamino nitrosobenzene, benzenamine, n,n-dimethyl-4-nitroso | |
| CHEBI:59990 | |
| CN(C)C1=CC=C(C=C1)N=O |
Specifications
| 138-89-6 | |
| 1.15 | |
| MFCD00002063 | |
| UN1369 | |
| 14,6638 | |
| CMEWLCATCRTSGF-UHFFFAOYSA-N | |
| N,N-dimethyl-4-nitrosoaniline | |
| 8749 | |
| 150.18 | |
| N,N-Dimethyl-4-nitrosoaniline |
| 84°C to 87°C | |
| C8H10N2O | |
| 25 g | |
| 607293 | |
| ndma, accelerine, p-nitroso-n,n-dimethylaniline, n,n-dimethyl-p-nitrosoaniline, 4-nitroso-n,n-dimethylaniline, 4-nitrosodimethylaniline, p-nitrosodimethylanilide, paranitrosodimethylanilide, 4-dimethylamino nitrosobenzene, benzenamine, n,n-dimethyl-4-nitroso | |
| CN(C)C1=CC=C(C=C1)N=O | |
| 150.181 | |
| CHEBI:59990 | |
| 98% |
Safety and Handling
GHS H Statement
H228-H251-H301-H310-H351-H373
Flammable solid.
Self-heating: may catch fire.
Toxic if swallowed.
Fatal in contact with skin.
Suspected of causing cancer.
May cause damage to organs through prolonged or repeated exposure.
P210-P235+P410-P260-P262-P264b-P270-P271-P280-P301+P310-P302+P352-P304+P340-P311-P312-P314-P330-P362-P370+P380-P380-P501c
H301+H311+H331-H373-H500-X
DOTInformation : Transport Hazard Class: 4.2; Packing Group: II; Proper Shipping Name: P-NITROSODIMETHYLANILINE
EINECSNumber : 205-343-1
RTECSNumber : BX7175000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only