missing translation for 'onlineSavingsMsg'
Learn More

N,N-Dimethyl-4-nitrosoaniline, 98%

Catalog No. AAA1830114
Change view
Click to view available options
Quantity:
25 g
250 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1830114 25 g
AAA1830130 250 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1830114 Supplier Thermo Scientific Chemicals Supplier No. A1830114

CAS: 138-89-6 | C8H10N2O | 150.181 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 138-89-6
Molecular Formula C8H10N2O
Molecular Weight (g/mol) 150.181
MDL Number MFCD00002063
InChI Key CMEWLCATCRTSGF-UHFFFAOYSA-N
Synonym ndma, accelerine, p-nitroso-n,n-dimethylaniline, n,n-dimethyl-p-nitrosoaniline, 4-nitroso-n,n-dimethylaniline, 4-nitrosodimethylaniline, p-nitrosodimethylanilide, paranitrosodimethylanilide, 4-dimethylamino nitrosobenzene, benzenamine, n,n-dimethyl-4-nitroso
PubChem CID 8749
ChEBI CHEBI:59990
IUPAC Name N,N-dimethyl-4-nitrosoaniline
SMILES CN(C)C1=CC=C(C=C1)N=O

Specifications

Melting Point 84°C to 87°C
Density 1.15
Quantity 25 g
UN Number UN1369
Beilstein 607293
Merck Index 14,6638
Formula Weight 150.18
Percent Purity 98%
Chemical Name or Material N,N-Dimethyl-4-nitrosoaniline
Hazard Category H301+H311+H331-H373-H500-X
Hazard Statement GHS H Statement
H228-H251-H301-H310-H351-H373
Flammable solid.
Self-heating: may catch fire.
Toxic if swallowed.
Fatal in contact with skin.
Suspected of causing cancer.
May cause damage to organs through prolonged or repeated exposure.
Precautionary Statement P210-P235+P410-P260-P262-P264b-P270-P271-P280-P301+P310-P302+P352-P304+P340-P311-P312-P314-P330-P362-P370+P380-P380-P501c
DOTInformation Transport Hazard Class: 4.2; Packing Group: II; Proper Shipping Name: P-NITROSODIMETHYLANILINE
EINECSNumber 205-343-1
RTECSNumber BX7175000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only