missing translation for 'onlineSavingsMsg'
Learn More

N,N-Dimethyl-4-nitroaniline 98.0+%, TCI America™

Catalog No. p-7126415
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D07605G 5 g
D076025G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D07605G Supplier TCI America Supplier No. D07605G
Only null left

Chemical Identifiers

CAS 100-23-2
Molecular Formula C8H10N2O2
Molecular Weight (g/mol) 166.18
MDL Number MFCD00014712
InChI Key QJAIOCKFIORVFU-UHFFFAOYSA-N
Synonym 4-nitro-n,n-dimethylaniline, benzenamine, n,n-dimethyl-4-nitro, 4-nitrodimethylaniline, p-nitro-n,n-dimethylaniline, p-nitrodimethylaniline, 1-dimethylamino-4-nitrobenzene, 4-dimethylamino nitrobenzene, p-dimethylamino nitrobenzene, n,n-dimethyl-p-nitroaniline, aniline, n,n-dimethyl-p-nitro
PubChem CID 7491
IUPAC Name N,N-dimethyl-4-nitroaniline
SMILES CN(C)C1=CC=C(C=C1)[N+](=O)[O-]

Specifications

Melting Point 165°C
Quantity 5 g
Formula Weight 166.18
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material N,N-Dimethyl-4-nitroaniline
RTECSNumber BX7035000
TSCA Yes