missing translation for 'onlineSavingsMsg'
Learn More

N,N-Dimethyl-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)aniline 98.0+%, TCI America™

Catalog No. D38321G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D38321G 1 g
D38325G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D38321G Supplier TCI America Supplier No. D38321G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 171364-78-6
Molecular Formula C14H22BNO2
Molecular Weight (g/mol) 247.15
MDL Number MFCD05663854
InChI Key DGMLJJIUOFKPKB-UHFFFAOYSA-N
Synonym n,n-dimethyl-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl aniline, 4-n,n-dimethylamino phenylboronic acid, pinacol ester, dimethyl 4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl phenyl amine, 4-dimethylamino phenylboronic acid pinacol ester, 4-n,n-dimethylamino phenylboronic acid pinacol ester, 2-4-dimethylamino phenyl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, n,n-dimethyl 4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl phenyl amine, n,n-dimethyl-4-tetramethyl-1,3,2-dioxaborolan-2-yl aniline, acmc-209e2t, 4-n,n-dimethylamino phenylboronic acid,pinacol ester
PubChem CID 2758659
IUPAC Name N,N-dimethyl-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)aniline
SMILES CN(C)C1=CC=C(C=C1)B1OC(C)(C)C(C)(C)O1

Specifications

Melting Point 125°C
Color White-Yellow
Quantity 1 g
Formula Weight 247.15
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material N,N-Dimethyl-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)aniline