Learn More
N,N-Dimethyl-3-nitroaniline, 98%, Thermo Scientific™
Supplier: Thermo Scientific Chemicals 116470250
| Quantity | 25g |
|---|---|
| Packaging | Glass bottle |
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Chemical Identifiers
| C8H10N2O2 | |
| MFCD00007236 | |
| 3-nitro-n,n-dimethylaniline, benzenamine, n,n-dimethyl-3-nitro, m-nitro-n,n-dimethylaniline, n,n-dimethyl-m-nitroaniline, 1-dimethylamino-3-nitrobenzene, benzene, 1-dimethylamino-3-nitro, aniline, n,n-dimethyl-m-nitro, dimethyl-3-nitro-phenyl-amine, dimethyl 3-nitrophenyl amine, m-nitrodimethylaniline | |
| N,N-dimethyl-3-nitroaniline |
Specifications
| N, N-Dimethyl-3-nitroaniline | |
| 619-31-8 | |
| 100.0 | |
| 98% | |
| C8H10N2O2 | |
| 25g | |
| 12, 701 | |
| Solubility in water: insoluble | |
| CN(C)C1=CC(=CC=C1)[N+](=O)[O-] | |
| 166.18 | |
| 166.18 |
| 98% | |
| 97.5 | |
| Authentic | |
| Glass bottle | |
| O2NC6H4N(CH3)2 | |
| MFCD00007236 | |
| 3-nitro-n,n-dimethylaniline, benzenamine, n,n-dimethyl-3-nitro, m-nitro-n,n-dimethylaniline, n,n-dimethyl-m-nitroaniline, 1-dimethylamino-3-nitrobenzene, benzene, 1-dimethylamino-3-nitro, aniline, n,n-dimethyl-m-nitro, dimethyl-3-nitro-phenyl-amine, dimethyl 3-nitrophenyl amine, m-nitrodimethylaniline | |
| CJDICMLSLYHRPT-UHFFFAOYSA-N | |
| N,N-dimethyl-3-nitroaniline | |
| 69269 | |
| 98% |
Safety and Handling
GHS H Statement
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
GHS P Statement
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume
GHS Signal Word: Warning
EINECSNumber : 210-590-3
TSCA : TSCA