missing translation for 'onlineSavingsMsg'
Learn More

N,N'-Dimethyl-3,4,9,10-perylenetetracarboxylic Diimide 92.0+%, TCI America™

Catalog No. D44291G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D44291G 1 g
D44295G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D44291G Supplier TCI America Supplier No. D44291G

Chemical Identifiers

CAS 5521-31-3
Molecular Formula C26H14N2O4
Molecular Weight (g/mol) 418.408
MDL Number MFCD00071975
InChI Key PJQYNUFEEZFYIS-UHFFFAOYSA-N
Synonym Pigment Red 179
PubChem CID 79657
SMILES CN1C(=O)C2=C3C(=CC=C4C3=C(C=C2)C5=C6C4=CC=C7C6=C(C=C5)C(=O)N(C7=O)C)C1=O

Specifications

Quantity 1 g
Formula Weight 418.41
Percent Purity ≥92.0% (N)
Physical Form Crystalline Powder
Chemical Name or Material N,N′-Dimethyl-3,4,9,10-perylenetetracarboxylic Diimide
EINECSNumber (5)-1112&(5)-3278
RTECSNumber CB1590000
TSCA Yes