missing translation for 'onlineSavingsMsg'
Learn More

N,N-Dimethyl-1H-imidazole-1-sulfonamide 98.0+%, TCI America™

Catalog No. p-7128771
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D54371G 1 g
D54375G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D54371G Supplier TCI America Supplier No. D54371G

Chemical Identifiers

CAS 78162-58-0
Molecular Formula C5H9N3O2S
Molecular Weight (g/mol) 175.206
MDL Number MFCD00955713
InChI Key YRRWNBMOJMMXQY-UHFFFAOYSA-N
Synonym 1-(Dimethylsulfamoyl)-1H-imidazole
PubChem CID 395751
IUPAC Name N,N-dimethylimidazole-1-sulfonamide
SMILES CN(C)S(=O)(=O)N1C=CN=C1

Specifications

Melting Point 49°C
Color White-Yellow
Boiling Point 100°C
Quantity 1 g
Formula Weight 175.21
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material N,N-Dimethyl-1H-imidazole-1-sulfonamide
Recommended Storage Refrigerator