missing translation for 'onlineSavingsMsg'
Learn More
Learn More
N,N'-Dimethyl-1,6-diaminohexane 97.0+%, TCI America™
$281.48 - $594.63
Chemical Identifiers
| CAS | 13093-04-4 |
|---|---|
| Molecular Formula | C8H22N2 |
| Molecular Weight (g/mol) | 146.28 |
| MDL Number | MFCD00008293 |
| InChI Key | MDKQJOKKKZNQDG-UHFFFAOYSA-P |
| PubChem CID | 83131 |
| IUPAC Name | methyl[6-(methylazaniumyl)hexyl]azanium |
| SMILES | C[NH2+]CCCCCC[NH2+]C |
Chemical Identifiers
| 13093-04-4 | |
| 146.28 | |
| MDKQJOKKKZNQDG-UHFFFAOYSA-P | |
| methyl[6-(methylazaniumyl)hexyl]azanium |
| C8H22N2 | |
| MFCD00008293 | |
| 83131 | |
| C[NH2+]CCCCCC[NH2+]C |
Specifications
| 13093-04-4 | |
| 205°C | |
| MFCD00008293 | |
| 2735 | |
| MDKQJOKKKZNQDG-UHFFFAOYSA-P | |
| methyl[6-(methylazaniumyl)hexyl]azanium | |
| 83131 | |
| ≥97.0% (GC) | |
| N,N′-Dimethyl-1,6-diaminohexane |
| Yellow | |
| C8H22N2 | |
| 10 mL | |
| 1,6-Bis(methylamino)hexane, N,N′C-Dimethyl-1,6-hexanediamine | |
| C[NH2+]CCCCCC[NH2+]C | |
| 146.28 | |
| 144.26 | |
| Liquid |
Safety and Handling
TSCA : Yes