missing translation for 'onlineSavingsMsg'
Learn More

N,N-Dimethyl-1,4-phenylenediamine Sulfate 98.0+%, TCI America™

Catalog No. p-7126428
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D078225G 25 g
D0782500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D078225G Supplier TCI America Supplier No. D078225G

Chemical Identifiers

CAS 536-47-0
Molecular Formula C8H14N2O4S
Molecular Weight (g/mol) 234.27
MDL Number MFCD00012992
InChI Key GLUKPDKNLKRLHX-UHFFFAOYSA-N
Synonym n,n-dimethyl-1,4-phenylenediamine sulfate, n,n-dimethyl-p-phenylenediamine sulfate, n1,n1-dimethylbenzene-1,4-diamine sulfate, 1,4-benzenediamine, n,n-dimethyl-, sulfate, unii-x997ivp3ja, 4-amino-n,n-dimethylaniline sulphate, x997ivp3ja, 1,4-benzenediamine, n,n-dimethyl-, sulfate 1:1, dimethyl-p-phenylenediamine; sulfuric acid, 1,4-benzenediamine, n1,n1-dimethyl-, sulfate 1:?
PubChem CID 80351
IUPAC Name N1,N1-dimethylbenzene-1,4-diamine; sulfuric acid
SMILES OS(O)(=O)=O.CN(C)C1=CC=C(N)C=C1

Specifications

Melting Point 205°C
Color White
Quantity 25 g
UN Number 2811
Formula Weight 234.27
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material N,N-Dimethyl-1,4-phenylenediamine Sulfate
EINECSNumber (3)-0243
RTECSNumber ST1575000
TSCA Yes
Recommended Storage Refrigerator