missing translation for 'onlineSavingsMsg'
Learn More
Learn More
N,N-Dimethyl-1,4-phenylenediamine Oxalate 98.0+%, TCI America™
Specifications
Specifications
| Quantity | 25 g |
| UN Number | 2811 |
| Formula Weight | 362.43 |
| Percent Purity | ≥98.0% (T) |
| Physical Form | Crystalline Powder |
| Chemical Name or Material | N,N-Dimethyl-1,4-phenylenediamine Oxalate |
Safety and Handling
| EINECSNumber | (3)-0243 |
| RTECSNumber | CZ1586550 |
| TSCA | Yes |