missing translation for 'onlineSavingsMsg'
Learn More

N,N-Dimethyl-1,4-phenylenediamine Dihydrochloride 99.0+%, TCI America™

Catalog No. D39315G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D39315G 5 g
D39311G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D39315G Supplier TCI America Supplier No. D39315G

Chemical Identifiers

CAS 536-46-9
Molecular Formula C8H14Cl2N2
Molecular Weight (g/mol) 209.11
MDL Number MFCD00012991
InChI Key IAEDWDXMFDKWFU-UHFFFAOYSA-N
Synonym n,n-dimethyl-p-phenylenediamine dihydrochloride, 4-amino-n,n-dimethylaniline dihydrochloride, n,n-dimethyl-1,4-phenylenediamine dihydrochloride, 1,4-benzenediamine, n,n-dimethyl-, dihydrochloride, 1,4-amino-n,n-dimethylaniline,dihydrochloride, n,n-dimethyl-1,4-benzenediamine dihydrochloride, n,n-dimethyl-p-phenylenediamine 2hcl, dimethyl-p-phenylenediamine dihydrochloride, n,n-dimethyl-p-phenylenediammonium dichloride, pubchem21651
PubChem CID 2724166
IUPAC Name N1,N1-dimethylbenzene-1,4-diamine dihydrochloride
SMILES Cl.Cl.CN(C)C1=CC=C(N)C=C1

Specifications

Melting Point 208°C
Quantity 5 g
UN Number 2811
Formula Weight 209.11
Percent Purity ≥99.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material N,N-Dimethyl-1,4-phenylenediamine Dihydrochloride [for Biochemical Research]
EINECSNumber (3)-0243
RTECSNumber ST1300000
TSCA Yes
Recommended Storage Refrigerator