missing translation for 'onlineSavingsMsg'
Learn More
Learn More
N,N-Dimethyl-1,4-butanediamine, 98%
Description
N,N-Dimethyl-1,4-butanediamine is used as pharmaceutical intermediate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
N,N-Dimethyl-1,4-butanediamine is used as pharmaceutical intermediate.
Notes
Air sensitive. Store away from oxidizing agents.
N,N-Dimethyl-1,4-butanediamine is used as pharmaceutical intermediate.
Notes
Air sensitive. Store away from oxidizing agents.
Specifications
Specifications
| Refractive Index | 1.44 |
| Linear Formula | (CH3)2NCH2CH2CH2CH2NH2 |
| Quantity | 5 g |
| UN Number | UN2735 |
| Sensitivity | Air Sensitive |
| Formula Weight | 116.21 |
| Percent Purity | 98% |
| Chemical Name or Material | N,N-Dimethyl-1,4-butanediamine |
Safety and Handling
| missing translation for 'healthHazard1' | H314-H335 |
| missing translation for 'healthHazard2' | GHS H Statement H314-H318 Causes severe skin burns and eye damage. Causes serious eye damage. |
| missing translation for 'healthHazard3' | P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c |
| DOTInformation | Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: AMINES, LIQUID, CORROSIVE, N.O.S. |
| EINECSNumber | 222-559-1 |
| TSCA | No |
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only