missing translation for 'onlineSavingsMsg'
Learn More

N,N-Dimethyl-1,4-butanediamine, 98%

Catalog No. AAH5027603
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH5027603 1 g
AAH5027606 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH5027603 Supplier Thermo Scientific Chemicals Supplier No. H5027603

CAS: 3529-10-0 | C6H16N2 | 116.208 g/mol

N,N-Dimethyl-1,4-butanediamine is used as pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
N,N-Dimethyl-1,4-butanediamine is used as pharmaceutical intermediate.

Notes
Air sensitive. Store away from oxidizing agents.

Chemical Identifiers

CAS 3529-10-0
Molecular Formula C6H16N2
Molecular Weight (g/mol) 116.208
MDL Number MFCD00047977
InChI Key GCOWZPRIMFGIDQ-UHFFFAOYSA-N
Synonym 4-dimethylaminobutylamine, 4-aminobutyl dimethylamine, 4-dimethylamino butylamine, n,n-dimethyl-1,4-butanediamine, n,n-dimethylbutane-1,4-diamine, n,n-dimethylaminobutylamine, chembl59625, nn-dimethyl-1,4-diaminobutane, 1,4-butanediamine, n,n-dimethyl, n1,n1-dimethyl-1,4-butanediamine
PubChem CID 77063
IUPAC Name N',N'-dimethylbutane-1,4-diamine
SMILES CN(C)CCCCN

Specifications

Refractive Index 1.44
Linear Formula (CH3)2NCH2CH2CH2CH2NH2
Quantity 1 g
UN Number UN2735
Sensitivity Air Sensitive
Formula Weight 116.21
Percent Purity 98%
Chemical Name or Material N,N-Dimethyl-1,4-butanediamine
Hazard Category H314-H335
Hazard Statement GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
Precautionary Statement P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
DOTInformation Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: AMINES, LIQUID, CORROSIVE, N.O.S.
EINECSNumber 222-559-1
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only