missing translation for 'onlineSavingsMsg'
Learn More

N,N-Dimethyl-1,3-cyclohexanediamine (cis- and trans- mixture) 98.0+%, TCI America™

Catalog No. D47401G
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D47401G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D47401G Supplier TCI America Supplier No. D47401G

Chemical Identifiers

CAS 885280-64-8
Molecular Formula C8H18N2
Molecular Weight (g/mol) 142.246
MDL Number MFCD06658433
InChI Key KKGNWXHXFXNADS-UHFFFAOYSA-N
Synonym 1-Amino-3-(dimethylamino)cyclohexane
PubChem CID 53249858
IUPAC Name 1-N,1-N-dimethylcyclohexane-1,3-diamine
SMILES CN(C)C1CCCC(C1)N

Specifications

Color Colorless
Quantity 1 g
UN Number 2735
Formula Weight 142.25
Percent Purity ≥98.0% (GC,T)
Physical Form Liquid
Chemical Name or Material N,N-Dimethyl-1,3-cyclohexanediamine (cis- and trans- mixture)