missing translation for 'onlineSavingsMsg'
Learn More

N,N'-Dimethoxy-N,N'-dimethyloxamide 98.0+%, TCI America™

Catalog No. D29615G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D29615G 5 g
D296125G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D29615G Supplier TCI America Supplier No. D29615G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 106675-70-1
Molecular Formula C6H12N2O4
Molecular Weight (g/mol) 176.172
MDL Number MFCD02326951
InChI Key OXSWGKZLVLVHIS-UHFFFAOYSA-N
PubChem CID 3287678
IUPAC Name N,N'-dimethoxy-N,N'-dimethyloxamide
SMILES CN(C(=O)C(=O)N(C)OC)OC

Specifications

Melting Point 93°C
Color White
Quantity 5 g
Synonym N,N′C-Oxalylbis[methoxy(methyl)amine]
Formula Weight 176.17
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material N,N′-Dimethoxy-N,N′-dimethyloxamide