missing translation for 'onlineSavingsMsg'
Learn More

N,N-Diisopropylformamide 98.0+%, TCI America™

$50.05 - $198.30

Chemical Identifiers

CAS 2700-30-3
Molecular Formula C7H15NO
Molecular Weight (g/mol) 129.203
MDL Number MFCD00008867
InChI Key UNBDDZDKBWPHAX-UHFFFAOYSA-N
Synonym n,n-diisopropylformamide, formamide, n,n-bis 1-methylethyl, n-formyldiisopropylamine, diisopropylformamide, n,n-bis 1-methylethyl formamide, nn-di-iso-propylformamide, formamide, n,n-diisopropyl, formamide, bis 1-methylethyl, n,n-bis methylethyl carboxamide, 1y1&nvhy1&1
PubChem CID 75912
IUPAC Name N,N-di(propan-2-yl)formamide
SMILES CC(C)N(C=O)C(C)C
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
D16515ML
View Documents
TCI America
D16515ML
5 mL
Each for $50.05
Only null left
 
D165125ML
View Documents
TCI America
D165125ML
25 mL
Each for $198.30
Only null left
 
Description

Description

Specifications

Chemical Identifiers

2700-30-3
129.203
UNBDDZDKBWPHAX-UHFFFAOYSA-N
75912
CC(C)N(C=O)C(C)C
C7H15NO
MFCD00008867
n,n-diisopropylformamide, formamide, n,n-bis 1-methylethyl, n-formyldiisopropylamine, diisopropylformamide, n,n-bis 1-methylethyl formamide, nn-di-iso-propylformamide, formamide, n,n-diisopropyl, formamide, bis 1-methylethyl, n,n-bis methylethyl carboxamide, 1y1&nvhy1&1
N,N-di(propan-2-yl)formamide

Specifications

2700-30-3
190°C
MFCD00008867
n,n-diisopropylformamide, formamide, n,n-bis 1-methylethyl, n-formyldiisopropylamine, diisopropylformamide, n,n-bis 1-methylethyl formamide, nn-di-iso-propylformamide, formamide, n,n-diisopropyl, formamide, bis 1-methylethyl, n,n-bis methylethyl carboxamide, 1y1&nvhy1&1
CC(C)N(C=O)C(C)C
129.203
129.20
Liquid
Yellow
C7H15NO
5 mL
UNBDDZDKBWPHAX-UHFFFAOYSA-N
N,N-di(propan-2-yl)formamide
75912
≥98.0% (GC)
N,N-Diisopropylformamide
Videos
Safety and Handling

Safety and Handling

TSCA : No

SDS
missing translation for 'documents'

missing translation for 'documents'