missing translation for 'onlineSavingsMsg'
Learn More
Learn More
N,N-Diisopropylformamide 98.0+%, TCI America™
Specifications
Specifications
| Color | Yellow |
| Boiling Point | 190°C |
| Quantity | 5 mL |
| Formula Weight | 129.20 |
| Percent Purity | ≥98.0% (GC) |
| Physical Form | Liquid |
| Chemical Name or Material | N,N-Diisopropylformamide |
Safety and Handling
| TSCA | No |