missing translation for 'onlineSavingsMsg'
Learn More

N,N-Diisopropylformamide, 98%

CAS: 2700-30-3 | C7H15NO | 129.203 g/mol

$87.74 - $328.34

Chemical Identifiers

CAS 2700-30-3
Molecular Formula C7H15NO
Molecular Weight (g/mol) 129.203
MDL Number MFCD00008867
InChI Key UNBDDZDKBWPHAX-UHFFFAOYSA-N
Synonym n,n-diisopropylformamide, formamide, n,n-bis 1-methylethyl, n-formyldiisopropylamine, diisopropylformamide, n,n-bis 1-methylethyl formamide, nn-di-iso-propylformamide, formamide, n,n-diisopropyl, formamide, bis 1-methylethyl, n,n-bis methylethyl carboxamide, 1y1&nvhy1&1
PubChem CID 75912
IUPAC Name N,N-di(propan-2-yl)formamide
SMILES CC(C)N(C=O)C(C)C
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAL0885706
View Documents
Thermo Scientific Chemicals
L0885706
5 g
Each for $87.74
Only null left
 
AAL0885714
View Documents
Thermo Scientific Chemicals
L0885714
25 g
Each for $328.34
Only null left
 
Description

Description

It is employed as a intermediate for organic synthesis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is employed as a intermediate for organic synthesis.

Solubility
Slightly soluble in water.

Notes
Store in cool. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.
Specifications

Chemical Identifiers

2700-30-3
129.203
UNBDDZDKBWPHAX-UHFFFAOYSA-N
75912
CC(C)N(C=O)C(C)C
C7H15NO
MFCD00008867
n,n-diisopropylformamide, formamide, n,n-bis 1-methylethyl, n-formyldiisopropylamine, diisopropylformamide, n,n-bis 1-methylethyl formamide, nn-di-iso-propylformamide, formamide, n,n-diisopropyl, formamide, bis 1-methylethyl, n,n-bis methylethyl carboxamide, 1y1&nvhy1&1
N,N-di(propan-2-yl)formamide

Specifications

2700-30-3
0.889
75°C (167°F)
1.437
5 g
n,n-diisopropylformamide, formamide, n,n-bis 1-methylethyl, n-formyldiisopropylamine, diisopropylformamide, n,n-bis 1-methylethyl formamide, nn-di-iso-propylformamide, formamide, n,n-diisopropyl, formamide, bis 1-methylethyl, n,n-bis methylethyl carboxamide, 1y1&nvhy1&1
UNBDDZDKBWPHAX-UHFFFAOYSA-N
N,N-di(propan-2-yl)formamide
75912
98%
10°C to 11°C
195°C to 196°C
C7H15NO
MFCD00008867
1745406
Slightly soluble in water.
CC(C)N(C=O)C(C)C
129.203
129.2
N,N-Diisopropylformamide
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H227
Combustible liquid.

P210-P235-P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P370+P378q-P501c

H227-H315-H319-H335

EINECSNumber : 220-284-1

TSCA : No

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only