missing translation for 'onlineSavingsMsg'
Learn More

N,N-Diisopropylformamide, 98%

Catalog No. AAL0885714
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0885714 25 g
AAL0885706 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL0885714 Supplier Thermo Scientific Chemicals Supplier No. L0885714

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 2700-30-3 | C7H15NO | 129.203 g/mol

It is employed as a intermediate for organic synthesis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is employed as a intermediate for organic synthesis.

Solubility
Slightly soluble in water.

Notes
Store in cool. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.

Chemical Identifiers

CAS 2700-30-3
Molecular Formula C7H15NO
Molecular Weight (g/mol) 129.203
MDL Number MFCD00008867
InChI Key UNBDDZDKBWPHAX-UHFFFAOYSA-N
Synonym n,n-diisopropylformamide, formamide, n,n-bis 1-methylethyl, n-formyldiisopropylamine, diisopropylformamide, n,n-bis 1-methylethyl formamide, nn-di-iso-propylformamide, formamide, n,n-diisopropyl, formamide, bis 1-methylethyl, n,n-bis methylethyl carboxamide, 1y1&nvhy1&1
PubChem CID 75912
IUPAC Name N,N-di(propan-2-yl)formamide
SMILES CC(C)N(C=O)C(C)C

Specifications

Melting Point 10°C to 11°C
Density 0.889
Boiling Point 195°C to 196°C
Flash Point 75°C (167°F)
Refractive Index 1.437
Quantity 25 g
Beilstein 1745406
Solubility Information Slightly soluble in water.
Formula Weight 129.2
Percent Purity 98%
Chemical Name or Material N,N-Diisopropylformamide
Show More Show Less
Hazard Category H227-H315-H319-H335
Hazard Statement GHS H Statement
H227
Combustible liquid.
Precautionary Statement P210-P235-P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P370+P378q-P501c
EINECSNumber 220-284-1
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only