missing translation for 'onlineSavingsMsg'
Learn More
Learn More
N,N'-Diisopropylethylenediamine 97.0+%, TCI America™
$82.79 - $246.43
Chemical Identifiers
| CAS | 4013-94-9 |
|---|---|
| Molecular Formula | C8H20N2 |
| Molecular Weight (g/mol) | 144.26 |
| MDL Number | MFCD00008864 |
| InChI Key | MFIGJRRHGZYPDD-UHFFFAOYSA-N |
| Synonym | n,n'-diisopropylethylenediamine, 1,2-ethanediamine, n,n'-bis 1-methylethyl, 1,2-ethanediamine, n1,n2-bis 1-methylethyl, n,n'-diisopropylethylene diamine, n,n'-di propan-2-yl ethane-1,2-diamine, 1,2-bis isopropylamino ethane, n,n'-disopropylethylene diamine, 1,2-ethanediamine,n1,n2-dipropyl, diped, n,n'-diisopropyl-ethylenediamine |
| PubChem CID | 77628 |
| IUPAC Name | (propan-2-yl)({2-[(propan-2-yl)amino]ethyl})amine |
| SMILES | CC(C)NCCNC(C)C |
Chemical Identifiers
| 4013-94-9 | |
| 144.26 | |
| MFIGJRRHGZYPDD-UHFFFAOYSA-N | |
| 77628 | |
| CC(C)NCCNC(C)C |
| C8H20N2 | |
| MFCD00008864 | |
| n,n'-diisopropylethylenediamine, 1,2-ethanediamine, n,n'-bis 1-methylethyl, 1,2-ethanediamine, n1,n2-bis 1-methylethyl, n,n'-diisopropylethylene diamine, n,n'-di propan-2-yl ethane-1,2-diamine, 1,2-bis isopropylamino ethane, n,n'-disopropylethylene diamine, 1,2-ethanediamine,n1,n2-dipropyl, diped, n,n'-diisopropyl-ethylenediamine | |
| (propan-2-yl)({2-[(propan-2-yl)amino]ethyl})amine |
Specifications
| 4013-94-9 | |
| 170°C | |
| MFCD00008864 | |
| 2733 | |
| MFIGJRRHGZYPDD-UHFFFAOYSA-N | |
| (propan-2-yl)({2-[(propan-2-yl)amino]ethyl})amine | |
| 77628 | |
| ≥97.0% (GC,T) | |
| N,N′-Diisopropylethylenediamine |
| Yellow | |
| C8H20N2 | |
| 5 g | |
| n,n'-diisopropylethylenediamine, 1,2-ethanediamine, n,n'-bis 1-methylethyl, 1,2-ethanediamine, n1,n2-bis 1-methylethyl, n,n'-diisopropylethylene diamine, n,n'-di propan-2-yl ethane-1,2-diamine, 1,2-bis isopropylamino ethane, n,n'-disopropylethylene diamine, 1,2-ethanediamine,n1,n2-dipropyl, diped, n,n'-diisopropyl-ethylenediamine | |
| CC(C)NCCNC(C)C | |
| 144.26 | |
| 144.26 | |
| Liquid |
Safety and Handling
TSCA : Yes