missing translation for 'onlineSavingsMsg'
Learn More

N,N'-Diisopropylethylenediamine 97.0+%, TCI America™

Catalog No. D39235G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D39235G 5 g
D392325G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D39235G Supplier TCI America Supplier No. D39235G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 4013-94-9
Molecular Formula C8H20N2
Molecular Weight (g/mol) 144.26
MDL Number MFCD00008864
InChI Key MFIGJRRHGZYPDD-UHFFFAOYSA-N
Synonym n,n'-diisopropylethylenediamine, 1,2-ethanediamine, n,n'-bis 1-methylethyl, 1,2-ethanediamine, n1,n2-bis 1-methylethyl, n,n'-diisopropylethylene diamine, n,n'-di propan-2-yl ethane-1,2-diamine, 1,2-bis isopropylamino ethane, n,n'-disopropylethylene diamine, 1,2-ethanediamine,n1,n2-dipropyl, diped, n,n'-diisopropyl-ethylenediamine
PubChem CID 77628
IUPAC Name (propan-2-yl)({2-[(propan-2-yl)amino]ethyl})amine
SMILES CC(C)NCCNC(C)C

Specifications

Color Yellow
Boiling Point 170°C
Quantity 5 g
UN Number 2733
Formula Weight 144.26
Percent Purity ≥97.0% (GC,T)
Physical Form Liquid
Chemical Name or Material N,N′-Diisopropylethylenediamine