missing translation for 'onlineSavingsMsg'
Learn More
Learn More
N,N-Diisopropylethylenediamine 97.0+%, TCI America™
Specifications
Specifications
| Color | Yellow |
| Boiling Point | 170°C |
| Quantity | 500 mL |
| UN Number | 2734 |
| Formula Weight | 144.26 |
| Percent Purity | ≥97.0% (GC,T) |
| Grade | Not Available |
| Physical Form | Liquid |
| Chemical Name or Material | N,N-Diisopropylethylenediamine |
Safety and Handling
| RTECSNumber | KV4200000 |
| TSCA | Yes |