missing translation for 'onlineSavingsMsg'
Learn More

N,N'-Diisopropylethylenediamine, 97%

Catalog No. AAA1711414
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1711414 25 g
AAA1711422 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1711414 Supplier Thermo Scientific Chemicals Supplier No. A1711414
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 4013-94-9 | C8H20N2 | 144.26 g/mol

N,N'-Diethylethylenediamine is used in the preparation of 2-imidazolidinecarboxaldehydes and 1,4,6,9-tetraalkyl-1,4,6,9-tetraaza-5,10-dioxaperhydroanthracenes by reacting with glyoxal. It acts as a ligand and form coordination complexes with copper(II), which shows thermochromic properties.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
N,N′-Diethylethylenediamine is used in the preparation of 2-imidazolidinecarboxaldehydes and 1,4,6,9-tetraalkyl-1,4,6,9-tetraaza-5,10-dioxaperhydroanthracenes by reacting with glyoxal. It acts as a ligand and form coordination complexes with copper(II), which shows thermochromic properties.

Solubility
Miscible with water.

Notes
Hygroscopic. Incompatible with strong oxidizing agents and strong acids.

Chemical Identifiers

CAS 4013-94-9
Molecular Formula C8H20N2
Molecular Weight (g/mol) 144.26
MDL Number MFCD00008864
InChI Key MFIGJRRHGZYPDD-UHFFFAOYSA-N
Synonym n,n'-diisopropylethylenediamine, 1,2-ethanediamine, n,n'-bis 1-methylethyl, 1,2-ethanediamine, n1,n2-bis 1-methylethyl, n,n'-diisopropylethylene diamine, n,n'-di propan-2-yl ethane-1,2-diamine, 1,2-bis isopropylamino ethane, n,n'-disopropylethylene diamine, 1,2-ethanediamine,n1,n2-dipropyl, diped, n,n'-diisopropyl-ethylenediamine
PubChem CID 77628
IUPAC Name N,N'-di(propan-2-yl)ethane-1,2-diamine
SMILES CC(C)NCCNC(C)C

Specifications

Density 0.798
Boiling Point 169°C to 171°C
Flash Point 51°C (123°F)
Refractive Index 1.4289
Quantity 25 g
UN Number UN2733
Beilstein 1735571
Sensitivity Hygroscopic
Solubility Information Miscible with water.
Formula Weight 144.26
Percent Purity 97%
Chemical Name or Material N,N'-Diisopropylethylenediamine
Show More Show Less
Hazard Category H226-H314-H335
Hazard Statement GHS H Statement
H314-H318-H226
Causes severe skin burns and eye damage.
Causes serious eye damage.
Flammable liquid and vapour.
Precautionary Statement P210-P233-P235-P240-P241-P242-P243-P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P370+P378q-P501c
DOTInformation Hazard Class: 8; Packaging Group: II
EINECSNumber 223-666-6
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only