missing translation for 'onlineSavingsMsg'
Learn More

N,N-Diisopropylethylamine trihydrofluoride, 90%, Thermo Scientific™

Catalog No. p-3559331
Change view
Click to view available options
Quantity:
1g
5g
Packaging:
Plastic bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC390710010 1g Plastic bottle
AC390710050 5g Plastic bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC390710010 Supplier Thermo Scientific Chemicals Supplier No. 390710010

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Chemical Identifiers

CAS 131600-43-6
Molecular Formula C8H22F3N
Molecular Weight (g/mol) 189.27
MDL Number MFCD00144880
InChI Key AJRRXKJZYYBJPY-UHFFFAOYSA-N
Synonym diisopropylethylamine trihydrofluoride, n-ethyl-n-isopropylpropan-2-amine trihydrofluoride, n,n-diisopropylethylamine trihydrofluoride, acmc-1bx1e, ksc490i4d, n,n,-diisopropylethylamine trihydrofluoride, n-ethyl-n-isopropylpropan-2-aminetrihydrofluoride, n-ethyl-n-isopropyl-propan-2-amine trihydrofluoride, n-ethyl-n-propan-2-yl-2-propanamine trihydrofluoride, n-ethyl-n-propan-2-yl-propan-2-amine trihydrofluoride
PubChem CID 45108321
IUPAC Name N-ethyl-N-propan-2-ylpropan-2-amine;trihydrofluoride
SMILES F.F.F.CCN(C(C)C)C(C)C

Specifications

CAS Min % 94.0
CAS Max % 100.0
Density 0.9650g/mL
Boiling Point 93°C to 95°C (4.0mmHg)
Flash Point 40.5°C
Infrared Spectrum Authentic
Assay Percent Range 90%
Packaging Plastic bottle
Quantity 1g
Specific Gravity 0.965
Solubility Information Solubility in water: soluble
Formula Weight 189.26
Percent Purity 90%
Chemical Name or Material N, N-Diisopropylethylamine trihydrofluoride
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Fatal in contact with skin.
Fatal if inhaled.
Fatal if swallowed.
Flammable liquid and vapor.
Causes severe skin burns and eye damage.
Precautionary Statement GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
IF ON SKIN (or hair): Take off immediately all contaminated clothing.
Rinse skin with water/shower.
Rinse mouth.
Immediately call a POISON CENT

RUO – Research Use Only