missing translation for 'onlineSavingsMsg'
Learn More

N,N-Diisopropylethylamine, 99+%

Catalog No. p-113599
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
2.5 kg
10 kg
Packaging:
Glass bottle
Plastic Bottle
Plastic Can
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC115220250 25 g Glass bottle
AC115221000 100 g Glass bottle
AC115225000 500 g Glass bottle
AC115220026 2.5 kg Plastic Bottle
AC115220101 10 kg Plastic Can
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. AC115220250 Supplier Thermo Scientific Chemicals Supplier No. 115220250
Only null left

CAS: 7087-68-5 | C8H19N | 129.24 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 7087-68-5
Molecular Formula C8H19N
Molecular Weight (g/mol) 129.24
MDL Number MFCD00008868
InChI Key JGFZNNIVVJXRND-UHFFFAOYSA-N
Synonym n,n-diisopropylethylamine, ethyldiisopropylamine, n-ethyldiisopropylamine, diisopropylethylamine, diea, hunig's base, n-ethyl-n-isopropylpropan-2-amine, dipea, 2-propanamine, n-ethyl-n-1-methylethyl, 1,1'-dimethyltriethylamine
PubChem CID 81531
IUPAC Name N-ethyl-N-propan-2-ylpropan-2-amine
SMILES CCN(C(C)C)C(C)C

Specifications

CAS Min % 98.0
CAS Max % 100.0
Melting Point -46.0°C
Color Colorless to Yellow
Density 0.7560g/mL
Boiling Point 127.0°C
Flash Point 6°C
Infrared Spectrum Authentic
Assay Percent Range 99% min. (GC)
Packaging Glass bottle
Refractive Index 1.4123 to 1.4143
Linear Formula [(CH3)2CH]2NC2H5
Quantity 25 g
Beilstein 04,IV,511
Fieser 01,371; 04,230; 07,148; 10,151; 12,277; 13,121; 16,176
Specific Gravity 0.756
Merck Index 15,3219
Solubility Information Solubility in water: 3.9g/L (20°C).
Viscosity 0.88 mm2/s (20°C)
Formula Weight 129.24
Percent Purity 99+%
Physical Form Liquid
Chemical Name or Material N,N-Diisopropylethylamine, 98+%
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Harmful if swallowed.
Toxic if inhaled.
Causes serious eye damage.
May cause respiratory irritation.
Precautionary Statement GHS P Statement
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if pre
EINECSNumber 230-392-0
TSCA TSCA

RUO – Research Use Only