missing translation for 'onlineSavingsMsg'
Learn More

N,N-Diisopropylethylamine, 99.5+%, AcroSeal™

Catalog No. p-7077808
Change view
Click to view available options
Quantity:
100 mL
1 L
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC459591000 100 mL
AC459590010 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC459591000 Supplier Thermo Scientific Chemicals Supplier No. 459591000
Only null left

CAS: 7087-68-5 | C8H19N | 129.24 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 7087-68-5
Molecular Formula C8H19N
Molecular Weight (g/mol) 129.24
InChI Key JGFZNNIVVJXRND-UHFFFAOYSA-N
Synonym n,n-diisopropylethylamine, ethyldiisopropylamine, n-ethyldiisopropylamine, diisopropylethylamine, diea, hunig's base, n-ethyl-n-isopropylpropan-2-amine, dipea, 2-propanamine, n-ethyl-n-1-methylethyl, 1,1'-dimethyltriethylamine
PubChem CID 81531
IUPAC Name N-ethyl-N-propan-2-ylpropan-2-amine
SMILES CCN(C(C)C)C(C)C

Specifications

Melting Point -46.0°C
Density 0.7560g/mL
Boiling Point 127.0°C
Flash Point 6°C
Infrared Spectrum Authentic
Assay Percent Range 99.5% min. (GC)
Refractive Index 1.4123 to 1.4143
Linear Formula [(CH3)2CH]2NC2H5
Quantity 100 mL
Specific Gravity 0.756
Solubility Information Solubility in water: 3.9g/L (20°C).
Viscosity 0.88 mm2/s (20°C)
Formula Weight 129.24
Percent Purity 99.5+%
Chemical Name or Material N,N-Diisopropylethylamine
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Harmful if swallowed.
Toxic if inhaled.
Causes serious eye damage.
May cause respiratory irritation.
Precautionary Statement GHS P Statement
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if pre
EINECSNumber 230-392-

RUO – Research Use Only