missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sigma Aldrich N,N-Diisopropylethylamine
MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.
$430.11 - $430.11
Specifications
| CAS | 7087-68-5 |
|---|---|
| Melting Point | <-50°C |
| Density | 0.742 g/mL at 25°C |
| Boiling Point | 127°C |
| Molecular Formula | C8H19N |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
387649100ML
|
Sigma Aldrich
387649100ML |
100 mL |
Each for $430.11
|
|
|||||
Description
N,N-DiisopropylethylamineSpecifications
| 7087-68-5 | |
| 0.742 g/mL at 25°C | |
| C8H19N | |
| n20/D 1.414 | |
| N-Ethyldiisopropylamine; DIPEA; Ethyldiisopropylamine; 'H++nig's base' |
| <-50°C | |
| 127°C | |
| [(CH3)2CH]2NC2H5 | |
| MFCD00008868 | |
| 129.24 g/mol |