missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sigma Aldrich N,N-Diisopropylethylamine
Description
N,N-Diisopropylethylamine
Spécifications
Spécifications
| CAS | 7087-68-5 |
| Melting Point | <-50°C |
| Density | 0.742 g/mL at 25°C |
| Boiling Point | 127°C |
| Molecular Formula | C8H19N |
| Refractive Index | n20/D 1.414 |
| Linear Formula | [(CH3)2CH]2NC2H5 |
| MDL Number | MFCD00008868 |
| Quantity | 100 mL |
| Synonym | N-Ethyldiisopropylamine; DIPEA; Ethyldiisopropylamine; 'H++nig's base' |
| Afficher plus de résultats |