Learn More
N,N'-Diisopropylcarbodiimide, 99%
Alfa Aesar N,N'-Diisopropylcarbodiimide, 99%, Quantity: 25g, Boiling Point: 145 deg.C to 148 deg.C, Molecular Weight: 126.203, Percent Purity: 99%, Beilstein: 878281, CAS: 693-13-0, ChEBI: CHEBI:53092, Density: 0.81, DOT Information: Hazard Class: 6.1; Packaging Group: I, EINECS Number: 211-743-7 | CAS: 693-13-0 | C7H14N2 | 126.203 g/mol
Supplier: Thermo Scientific Chemicals A1929230
Description
N,N'-Diisopropylcarbodiimide is used as a reagent in synthetic organic chemistry. It serves as a chemical intermediate and as a stabilizer for Sarin (chemical weapon). It is also used in the synthesis of peptide and nucleic acid. Further, it is used as an antineoplastic and involved in the treatment of malignant melanoma and sarcomas. In addition to this, it is used in the synthesis of acid anhydride, aldehyde, ketone and isocyanate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsN,N′-Diisopropylcarbodiimide is used as a reagent in synthetic organic chemistry. It serves as a chemical intermediate and as a stabilizer for Sarin (chemical weapon). It is also used in the synthesis of peptide and nucleic acid. Further, it is used as an antineoplastic and involved in the treatment of malignant melanoma and sarcomas. In addition to this, it is used in the synthesis of acid anhydride, aldehyde, ketone and isocyanate.
Solubility
Soluble in chloroform, methylene chloride, acetonitrile, dioxane, dimethylformamide andtetrahydrofuran.
Notes
Moisture sensitive. Store in a cool place Incompatible with oxidizing agents and acids.
Specifications
| N,N'-Diisopropylcarbodiimide | |
| 0.81 | |
| 33°C (91°F) | |
| C7H14N2 | |
| MFCD00065689 | |
| UN2929 | |
| Moisture sensitive | |
| Soluble in chloroform,methylene chloride,acetonitrile,dioxane,dimethylformamide andtetrahydrofuran. | |
| CC(C)N=C=NC(C)C | |
| 126.203 | |
| CHEBI:53092 | |
| 99% |
| 693-13-0 | |
| 145°C to 148°C | |
| Stench | |
| 1.433 | |
| 250 g | |
| 878281 | |
| n,n'-diisopropylcarbodiimide, diisopropylcarbodiimide, 1,3-diisopropylcarbodiimide, n,n-diisopropylcarbodiimide, carbodiimide, diisopropyl, dipcdi, dic, 2-propanamine, n,n'-methanetetraylbis, diisopropylmethanediimine, n,n'-methanediylidenebis propan-2-amine | |
| BDNKZNFMNDZQMI-UHFFFAOYSA-N | |
| N,N'-di(propan-2-yl)methanediimine | |
| 12734 | |
| 126.2 |
Chemical Identifiers
| 693-13-0 | |
| 126.203 | |
| BDNKZNFMNDZQMI-UHFFFAOYSA-N | |
| 12734 | |
| N,N'-di(propan-2-yl)methanediimine |
| C7H14N2 | |
| MFCD00065689 | |
| n,n'-diisopropylcarbodiimide, diisopropylcarbodiimide, 1,3-diisopropylcarbodiimide, n,n-diisopropylcarbodiimide, carbodiimide, diisopropyl, dipcdi, dic, 2-propanamine, n,n'-methanetetraylbis, diisopropylmethanediimine, n,n'-methanediylidenebis propan-2-amine | |
| CHEBI:53092 | |
| CC(C)N=C=NC(C)C |
Safety and Handling
GHS H Statement
H330-H334-H318-H226-H315-H317-H335
Fatal if inhaled.
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
Causes serious eye damage.
Flammable liquid and vapour.
Causes skin irritation.
May cause an allergic skin reaction.
May cause respiratory irritation.
P210-P233-P235-P240-P241-P242-P243-P260-P264b-P271-P272-P280-P284-P285-P303+P361+P353-P304+P340-P305+P351+P338-P310-P312-P333+P313-P363-P370+P378q-P501c
H226-H315-H317-H318-H330-H334-H335
DOTInformation : Hazard Class: 6.1; Packaging Group: I
EINECSNumber : 211-743-7
RTECSNumber : FF2175000
TSCA : Yes
Recommended Storage : Ambient temperatures; Store under Nitrogen
RUO – Research Use Only