missing translation for 'onlineSavingsMsg'
Learn More

N,N-Diisopropylcarbamoyl chloride, 98%

Catalog No. AC314520050
Change view
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC314520050 5 g Glass Bottle
AC314520250 25 g Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC314520050 Supplier Thermo Scientific Chemicals Supplier No. 314520050

CAS: 19009-39-3 | C7H14ClNO | 163.65 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 19009-39-3
Molecular Formula C7H14ClNO
Molecular Weight (g/mol) 163.65
InChI Key RSAFAYLZKCYUQW-UHFFFAOYSA-N
Synonym diisopropylcarbamoyl chloride, n,n-diisopropylcarbamoyl chloride, diisopropylcarbamic chloride, diisopropylcarbamyl chloride, n,n-di propan-2-yl carbamoyl chloride, acmc-20ak8x, diisopropyl carbamyl chloride, di-i-propylcarbamoyl chloride, n,n-diisopropylchloroformamide, n,n-diisopropylcarbamoylchloride
PubChem CID 87890
IUPAC Name N,N-di(propan-2-yl)carbamoyl chloride
SMILES CC(C)N(C(C)C)C(=O)Cl

Specifications

Melting Point 58.1°C to 59.1°C
Boiling Point 90.0°C to 93.0°C (15.0mmHg)
CAS Min % 97.5
CAS Max % 100.0
Color Colorless to Yellow or White to Yellow
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass Bottle
Linear Formula [(CH3)2CH]2NCOCl
Quantity 5 g
Formula Weight 163.65
Percent Purity 98%
Physical Form Crystalline Powder or Low Melting Solid
Chemical Name or Material N, N-Diisopropylcarbamoyl chloride
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Causes severe skin burns and eye damage.
Contact with water liberates toxic gas.
Precautionary Statement GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN (or hair): Take off im
EINECSNumber 242-743-5

RUO – Research Use Only