missing translation for 'onlineSavingsMsg'
Learn More

N,N'-Diisopropyl-O-methylisourea 98.0+%, TCI America™

Catalog No. D45875ML
Change view
Click to view available options
Quantity:
5 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D45875ML 5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D45875ML Supplier TCI America Supplier No. D45875ML

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 54648-79-2
Molecular Formula C8H18N2O
Molecular Weight (g/mol) 158.245
MDL Number MFCD00008866
InChI Key PUVRRPLSJKDMKH-UHFFFAOYSA-N
Synonym Methyl N,N′C-Diisopropylcarbamimidate, N,N′C-Diisopropylcarbamimidic Acid Methyl Ester
PubChem CID 383824
IUPAC Name methyl N,N'-di(propan-2-yl)carbamimidate
SMILES CC(C)NC(=NC(C)C)OC

Specifications

Color Colorless
Boiling Point 51°C
Quantity 5 mL
UN Number 1993
Formula Weight 158.25
Percent Purity ≥98.0% (T)
Physical Form Liquid
Chemical Name or Material N,N′-Diisopropyl-O-methylisourea
TSCA No
Recommended Storage Refrigerator