missing translation for 'onlineSavingsMsg'
Learn More

N,N'-Dihydroxypyromellitimide 96.0+%, TCI America™

$273.38 - $918.31

Chemical Identifiers

CAS 57583-53-6
Molecular Formula C10H4N2O6
Molecular Weight (g/mol) 248.15
InChI Key VBBUVIWLZBAEEF-UHFFFAOYSA-N
PubChem CID 221479
IUPAC Name 2,6-dihydroxypyrrolo[3,4-f]isoindole-1,3,5,7-tetrone
SMILES C1=C2C(=CC3=C1C(=O)N(C3=O)O)C(=O)N(C2=O)O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
D44131G
View Documents
TCI America
D44131G
1 g
Each for $273.38
Only null left
 
D44135G
View Documents
TCI America
D44135G
5 g
Each for $918.31
Only null left
 
Description

Description

Specifications

Chemical Identifiers

57583-53-6
248.15
221479
C1=C2C(=CC3=C1C(=O)N(C3=O)O)C(=O)N(C2=O)O
C10H4N2O6
VBBUVIWLZBAEEF-UHFFFAOYSA-N
2,6-dihydroxypyrrolo[3,4-f]isoindole-1,3,5,7-tetrone

Specifications

57583-53-6
C10H4N2O6
N,N′C-Dihydroxypyromellitic Diimide, NDHPI
C1=C2C(=CC3=C1C(=O)N(C3=O)O)C(=O)N(C2=O)O
248.15
248.15
Crystalline Powder
Yellow
1 g
VBBUVIWLZBAEEF-UHFFFAOYSA-N
2,6-dihydroxypyrrolo[3,4-f]isoindole-1,3,5,7-tetrone
221479
≥96.0% (HPLC,N)
N,N′-Dihydroxypyromellitimide
Videos
Safety and Handling

Safety and Handling

TSCA : No

SDS
missing translation for 'documents'

missing translation for 'documents'