missing translation for 'onlineSavingsMsg'
Learn More

N,N'-Dihydroxy-1,2,3,4-cyclobutanetetracarboxdiimide 98.0+%, TCI America™

Catalog No. d42121g
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D42121G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D42121G Supplier TCI America Supplier No. D42121G
Only null left

Chemical Identifiers

CAS 245049-70-1
Molecular Formula C8H6N2O6
Molecular Weight (g/mol) 226.144
InChI Key HONIHRUONDEYHT-UHFFFAOYSA-N
PubChem CID 68803161
SMILES C12C(C3C1C(=O)N(C3=O)O)C(=O)N(C2=O)O

Specifications

Color White-Yellow
Quantity 1 g
Formula Weight 226.14
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material N,N′-Dihydroxy-1,2,3,4-cyclobutanetetracarboxdiimide