missing translation for 'onlineSavingsMsg'
Learn More

N,N-Diethylnicotinamide 98.0+%, TCI America™

Catalog No. p-7126260
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D051425G 25 g
D0514500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D051425G Supplier TCI America Supplier No. D051425G

Chemical Identifiers

CAS 59-26-7
Molecular Formula C10H14N2O
Molecular Weight (g/mol) 178.235
MDL Number MFCD00006386
InChI Key NCYVXEGFNDZQCU-UHFFFAOYSA-N
Synonym nikethamide, n,n-diethylnicotinamide, nicethamide, corvin, anacardone, anacordone, carbamidal, coramine, corvotone, nicamide
PubChem CID 5497
IUPAC Name N,N-diethylpyridine-3-carboxamide
SMILES CCN(CC)C(=O)C1=CN=CC=C1

Specifications

Color White-Yellow
Boiling Point 300°C
Quantity 25 g
UN Number 2811
Formula Weight 178.24
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Lumps
Chemical Name or Material N,N-Diethylnicotinamide
EINECSNumber (5)-0739
RTECSNumber QS4375000
TSCA No