missing translation for 'onlineSavingsMsg'
Learn More

N,N-Diethylmethylamine 98.0+%, TCI America™

Catalog No. p-7126611
Change view
Click to view available options
Quantity:
25 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D113825ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D113825ML Supplier TCI America Supplier No. D113825ML
Only null left

Chemical Identifiers

CAS 616-39-7
Molecular Formula C5H14N
Molecular Weight (g/mol) 88.17
MDL Number MFCD00009049
InChI Key GNVRJGIVDSQCOP-UHFFFAOYSA-O
Synonym n,n-diethylmethylamine, diethylmethylamine, ethanamine, n-ethyl-n-methyl, n-methyldiethylamine, diethylamine, n-methyl, n-ethyl-n-methyl-ethanamine, methyldiethylamine, diethylmethyl amine, diethyl methyl amine, methyl diethyl amine
PubChem CID 12022
IUPAC Name diethyl(methyl)azanium
SMILES CC[NH+](C)CC

Specifications

Color Yellow
Boiling Point 62°C
Quantity 25 mL
UN Number 2733
Formula Weight 87.17
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material N,N-Diethylmethylamine