missing translation for 'onlineSavingsMsg'
Learn More

N,N-Diethylhydroxylamine, 97%

Catalog No. p-109187
Change view
Click to view available options
Quantity:
100 g
500 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC114421000 100 g Glass bottle
AC114425000 500 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC114421000 Supplier Thermo Scientific Chemicals Supplier No. 114421000
Only null left

CAS: 3710-84-7 | C4H11NO | 89.14 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 3710-84-7
Molecular Formula C4H11NO
Molecular Weight (g/mol) 89.14
MDL Number MFCD00002126
InChI Key FVCOIAYSJZGECG-UHFFFAOYSA-N
Synonym diethylhydroxylamine, ethanamine, n-ethyl-n-hydroxy, n-hydroxydiethylamine, n,n-diethylhydroxyamine, hydroxylamine, n,n-diethyl, unii-314i05edvh, ccris 964, hydroxydiethylamine, diethyl hydroxylamine, diethyl hydroxykamine
PubChem CID 19463
IUPAC Name N,N-diethylhydroxylamine
SMILES CCN(CC)O

Specifications

CAS Min % 96.0
CAS Max % 100.0
Melting Point -26°C to 25°C
Density 0.8670g/mL
Boiling Point 125.0°C to 130.0°C
Flash Point 45°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Refractive Index 1.4190 to 1.4220
Linear Formula (C2H5)2NOH
Quantity 100 g
Beilstein 04, 536
Fieser 06,187
Specific Gravity 0.867
Solubility Information Solubility in water: soluble.
Formula Weight 89.14
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material N, N-Diethylhydroxylamine, 97%
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Flammable liquid and vapor.
Harmful in contact with skin.
Harmful if inhaled.
May cause respiratory irritation.
Toxic to aquatic life with long lasting effects.
Precautionary Statement GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fu
EINECSNumber 223-055-4
RTECSNumber NC3500000
TSCA TSCA

RUO – Research Use Only