missing translation for 'onlineSavingsMsg'
Learn More

N,N-Diethylhydroxylamine, 97%

Catalog No. AAB2330214
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2330214 25 g
AAB2330222 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAB2330214 Supplier Thermo Scientific Chemicals Supplier No. B2330214
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 3710-84-7 | C4H11NO | 89.138 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 3710-84-7
Molecular Formula C4H11NO
Molecular Weight (g/mol) 89.138
MDL Number MFCD00002126
InChI Key FVCOIAYSJZGECG-UHFFFAOYSA-N
Synonym diethylhydroxylamine, ethanamine, n-ethyl-n-hydroxy, n-hydroxydiethylamine, n,n-diethylhydroxyamine, hydroxylamine, n,n-diethyl, unii-314i05edvh, ccris 964, hydroxydiethylamine, diethyl hydroxylamine, diethyl hydroxykamine
PubChem CID 19463
IUPAC Name N,N-diethylhydroxylamine
SMILES CCN(CC)O

Specifications

Melting Point -25°C
Density 0.867
Boiling Point 125°C to 130°C
Flash Point 46°C (114°F)
Refractive Index 1.4215
Quantity 25 g
UN Number UN1993
Beilstein 1731349
Sensitivity Hygroscopic
Formula Weight 89.14
Percent Purity 97%
Chemical Name or Material N,N-Diethylhydroxylamine
Show More Show Less
Hazard Category H226-H312+H332-H335
Hazard Statement GHS H Statement
H311-H226-H341-H332-H315-H319-H335
Toxic in contact with skin.
Flammable liquid and vapour.
Suspected of causing genetic defects.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P210-P233-P235-P240-P241-P242-P243-P261-P271-P280-P303+P361+P353-P304+P340-P312-P363-P370+P378q-P501c
DOTInformation Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: FLAMMABLE LIQUIDS, N.O.S.
EINECSNumber 223-055-4
RTECSNumber NC3500000
TSCA Yes
Recommended Storage Ambient temperatures; Store under Nitrogen

RUO – Research Use Only