missing translation for 'onlineSavingsMsg'
Learn More

N,N-Diethylformamide 99.0+%, TCI America™

Catalog No. p-7126256
Change view
Click to view available options
Quantity:
25 mL
100 mL
500 mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D050625ML 25 mL
D0506100ML 100 mL
D0506500ML 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. D050625ML Supplier TCI America Supplier No. D050625ML
Only null left

Chemical Identifiers

CAS 617-84-5
Molecular Formula C5H11NO
Molecular Weight (g/mol) 101.15
MDL Number MFCD00003287
InChI Key SUAKHGWARZSWIH-UHFFFAOYSA-N
Synonym diethylformamide, formamide, n,n-diethyl, n-formyldiethylamine, diethyl formamide, formyldiethylamine, diethylamid kyseliny mravenci, n,n-diethylcarboxamide, diethylamid kyseliny mravenci czech, diethyl-formamide, formamide, diethyl
PubChem CID 12051
IUPAC Name N,N-diethylformamide
SMILES CCN(CC)C=O

Specifications

Color Yellow
Boiling Point 177°C
Quantity 25 mL
UN Number 1993
Formula Weight 101.15
Percent Purity ≥99.0% (GC)
Physical Form Liquid
Chemical Name or Material N,N-Diethylformamide
EINECSNumber (2)-3847
RTECSNumber LQ1925000
TSCA Yes