missing translation for 'onlineSavingsMsg'
Learn More

N,N-Diethylformamide 99.0+%, TCI America™

Catalog No. d050625ml
Change view
missing translation for 'orderingAttributeHoverText'
Quantity:
100 mL
25 mL
500 mL
Catalog No. Quantity
D050625ML 25 mL
D0506100ML 100 mL
D0506500ML 500 mL
3 options

Catalog No. D050625ML

Supplier: TCI America D050625ML

Only null left

Chemical Identifiers

CAS 617-84-5
Molecular Formula C5H11NO
Molecular Weight (g/mol) 101.15
MDL Number MFCD00003287
InChI Key SUAKHGWARZSWIH-UHFFFAOYSA-N
Synonym diethylformamide, formamide, n,n-diethyl, n-formyldiethylamine, diethyl formamide, formyldiethylamine, diethylamid kyseliny mravenci, n,n-diethylcarboxamide, diethylamid kyseliny mravenci czech, diethyl-formamide, formamide, diethyl
PubChem CID 12051
IUPAC Name N,N-diethylformamide
SMILES CCN(CC)C=O

Specifications

Color Yellow
Boiling Point 177°C
Quantity 25 mL
UN Number 1993
Formula Weight 101.15
Percent Purity ≥99.0% (GC)
Physical Form Liquid
Chemical Name or Material N,N-Diethylformamide
EINECSNumber (2)-3847
RTECSNumber LQ1925000
TSCA Yes