missing translation for 'onlineSavingsMsg'
Learn More

N,N-Diethylformamide 99.0+%, TCI America™

Catalog No. p-7128170
Change view
Click to view available options
Quantity:
100 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D4234100ML 100 mL
D4234500ML 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D4234100ML Supplier TCI America Supplier No. D4234100ML
Only null left

Chemical Identifiers

CAS 617-84-5
Molecular Formula C5H11NO
Molecular Weight (g/mol) 101.15
MDL Number MFCD00003287
InChI Key SUAKHGWARZSWIH-UHFFFAOYSA-N
Synonym diethylformamide, formamide, n,n-diethyl, n-formyldiethylamine, diethyl formamide, formyldiethylamine, diethylamid kyseliny mravenci, n,n-diethylcarboxamide, diethylamid kyseliny mravenci czech, diethyl-formamide, formamide, diethyl
PubChem CID 12051
IUPAC Name N,N-diethylformamide
SMILES CCN(CC)C=O

Specifications

Color Colorless
Boiling Point 177°C
Quantity 100 mL
UN Number 1993
Formula Weight 101.15
Percent Purity ≥99.0% (GC)
Physical Form Liquid
Chemical Name or Material N,N-Diethylformamide [for Biochemical Research]
EINECSNumber (2)-3847
RTECSNumber LQ1925000
TSCA Yes